Would you like to log in again?
KIMAX® Separatory Funnels, Squibb, Pear-Shaped, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
29048F-60 29048-60 29048-250 29049F-125 29048F-250 29048F-125 29048T-500 29048-500 29049F-1000 29048-1000 29048-125 29048F-1000 29049F-250 29048F-2000 29048T-125 29049F-2000 29048F-30 29048F-500 29049F-500 29048T-250 29048T-60 29048T-2000
89002-092CS 891.89 USD InStock
89002-092 89002-094 30352-142 89002-090 89002-088 89002-110 89002-122 89002-100 89002-112 89002-102 89002-124 89002-096 89002-120 89002-098 89002-118 89002-108 89002-114 89002-104 89002-126 89002-116 89002-106 89002-128
KIMAX® Separatory Funnels, Squibb, Pear-Shaped, Kimble Chase, DWK Life Sciences
Funnels Separating Funnels
Funnels are designed with a large lower stem diameter to allow more complete separation.
  • Pear-shaped
  • Autoclavable
Constructed with heavy walls, a fire-polished and chip-resistant stem, and heavily beaded rim.
Certifications: Designed from ASTM Specification E1096, Type 4 requirements. Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Supplied with a Standard Taper ground glass stopper and an autoclavable PTFE stopcock plug.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty