Would you like to log in again?
KIMAX® Centrifuge Tube, Conical-Bottom, Graduated, with Pourout, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
45186-50
89002-206CS 947.61 USD InStock
89002-206
KIMAX® Centrifuge Tube, Conical-Bottom, Graduated, with Pourout, Kimble Chase
Tubes Centrifuge Tubes
Tube has a short tapered bottom and a pour spout.
  • Graduated and calibrated to contain
Scale, legend and marking spot are durable white ceramic enamel. Graduated to 10 ml in 0.5 ml increments and from 10 to 50 ml in 1 ml increments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty