Would you like to log in again?
KIMAX® Centrifuge Tubes, Graduated, Oil, with White Scales, Kimble Chase
Supplier: DWK Life Sciences
45244-100
89002-232CS 2363.55 USD InStock
89002-232
KIMAX® Centrifuge Tubes, Graduated, Oil, with White Scales, Kimble Chase
Tubes Centrifuge Tubes
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements and it is used in the determination of bottom sediment and water in petroleum products.
  • Calibrated to contain
  • Stem holds 1.5 ml
  • Scale and legend are durable white ceramic enamel
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty