Would you like to log in again?
PG2 DOT-Compliant Sharps Disposal Containers, Covidien
Supplier: Covidien
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8998PG2 8997PG2 8998PG2 8997PG2 8998SPG2 8930SA 8998SPG2 8936PG2
89002-264CS 442.35 USD InStock
89002-264 89002-266 76480-332 93000-308 CA89002-264 89004-532 CA89002-266 CA89004-532
PG2 DOT-Compliant Sharps Disposal Containers, Covidien
Bins Sharps Containers
These PG2-rated sharps disposal containers are ideal for the transport of sharps, cultures, and stocks
Container labeling is DOT-compliant, eliminating the need for secondary packaging when transporting by a private or contract carrier. Containers meet or exceed JCAHO, OSHA, and NIOSH/CDC standards and requirements.
An absorbent pad helps contain excess fluid. The lid gasket provides leak resistance. Molded carrying handles allow easy transport
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty