Would you like to log in again?
WYPALL® X80 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WYPALL®
05930 41041 41048 41026 41029 41029 41043 41048 41025 41055 41025 41044 41044 41055 41043 41041 05930 41026
89003-002CS 146.04 USD InStock
89003-002 CA89003-010 89003-010 CA89003-004 89003-842 89003-006 CA89003-002 89003-004 CA89003-008 CA89003-842 CA89003-006 89003-008 89000-634 CA89000-634 89004-384 CA89004-384 89004-556 CA89004-556
WYPALL® X80 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Reusable Cleaning Cloths Cloths for General Applications
Wipers absorb oil and water faster than rags or laundered towels.They maintain strength even in solvents. Long-lasting, on-the-job performance extends usage, reducing cost and waste.
  • Durable and economical alternative to laundered shop towels and rags
  • Suitable for a wide range of laboratory and industrial tasks, including oil, solvent, and chemical cleanup
  • Consistent size and strength without the inconvenience of holes, strings, or residues
  • HYDROKNIT® hydroentangled material manufactured without adhesives or chemical binders will not scratch skin or delicate surfaces
  • Reusable towels will not fall apart when wet
Ordering information: Quarter-fold packaged wipers are wrapped in stacks of 50. BRAG™ box-packaged wipers are supplied 160 per box; Pop-Up™ boxes each contain 80 wipers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty