Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
KIMAX® Glass Rods, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
80700-1 80700-8
89003-310CS 1003.73 USD InStock
89003-310 89003-324
KIMAX® Glass Rods, Kimble Chase, DWK Life Sciences
Stirrers Stirring Rods
Made of KG-33 borosilicate glass; which has a low coefficient of expansion
Ends are cut.
Certifications: Meets requirements for Type I Class A, ASTM E438.
Ordering information: All values are approximate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty