Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
KIMAX® Reusable Measuring (Mohr) Pipettes, Class B, Color-Coded, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
37020-1110 37020-11100 37020-50 37020-25 37020-5 37020-2 37020-10 37020-110
89003-508CS 253.25 USD InStock
89003-508 89003-506 72700-027 53103-191 89003-512 89003-510 89003-516 89003-514
KIMAX® Reusable Measuring (Mohr) Pipettes, Class B, Color-Coded, Kimble Chase, DWK Life Sciences
Pipettes Measuring (Mohr) Pipettes
Calibrated to deliver, these rugged KIMAX-51® pipettes are intended for chemical laboratory work.
  • Color coding facilitates sorting and selection
  • Designed from ASTM Specification E1293, Style 1, Class B requirements
Tip openings are smaller than typically desired for clinical laboratory applications. Scale is permanent brown stain fused into uniform bore tubing without etching. Pipettes are graduated to a base line which is on the straight tube above the taper.
Delivery information: Each pipette is supplied with a permanently marked individual serial number and a Certificate of Graduation Accuracy.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty