Would you like to log in again?
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
37034B-5 37034B-25 37034B-2 37034B-10
89003-528CS 354.32 USD InStock
89003-528 89003-530 89003-532 89003-526
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Pipettes Serological Pipettes
Wide-tip pipettes with large tip opening provide faster delivery and are ideal for use with viscous liquids, slurries, and suspensions
Made from KIMAX-51® borosilicate glass for the highest resistance to chemical attack and scratching. Pipettes are calibrated to deliver to the tip. Top ends are tooled to accept cotton plugging. Scale is permanent brown stain fused into the surface without etching. Pipettes are color-coded for ease in sorting and selection. Designed from ASTM specification E1044, Style III requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty