Would you like to log in again?
KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Screw Cap, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
60941A-12 60941A-8 60941A-16 60941A-24 60941A-40
11310-772CS 206.89 USD InStock
11310-772 11310-770 11310-768 89003-542 11310-776
KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Screw Cap, Kimble Chase, DWK Life Sciences
Vials
Clear, screw thread sample vials.
  • Black phenolic cap and cone-shaped polyethylene liner
  • Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Material conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty