Would you like to log in again?
Screw Thread Vials with Attached Closures, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
60942A-40 60942A-16 60942A-8 60942A-24
66010-230CS 284.66 USD InStock
66010-230 66010-234 66010-236 89003-544
Screw Thread Vials with Attached Closures, Kimble Chase, DWK Life Sciences
Vials
These glass, screw-thread sample vials are manufactured from 33 expansion, low extractable borosilicate glass confirming to USP Type I ASTM E438, Class A requirements.
Vials feature open-top, black, polypropylene closures with PTFE-faced, silicone septa attached. Vials are packed in corrugated trays with partitions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty