Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HANDI-VAC® Vacuum Handling System, Virtual Industries
Supplier: Virtual Industries
HANDI-VAC®
HV-X V3972B V9050-C V9038-C
89002-770EA 3.85 USD InStock
89002-770 89003-062 89003-588 89003-586
HANDI-VAC® Vacuum Handling System, Virtual Industries
Vacuum Handling Systems
This low-cost vaccum handling system works like a tweezer
Tool features a squeeze bulb design and is made of black static-dissipative rubber. Required vacuum cups are available in two sizes for various handling needs. Vaccum cups are constructed of clear high-temperature silicone and can attach directly to the tool or be expanded via the metal probe.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty