Would you like to log in again?
VWR® Microcentrifuge Tubes with Socket Screw-Cap
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
CFT474020 CFT481015 CFT484015 CFT473005 CFT485015 CFT471005 CFT474005 CFT470020 CFT472005 CFT470005 CFT471020 CFT480015 CFT483015 CFT472020 CFT482015 CFT475005 CFT473020 CFT476020
89004-308CS 1862.24 USD InStock
89004-308 89004-290 89004-280 89004-292 89004-282 89004-294 89004-284 89004-296 89004-286 89004-320 89004-298 89004-288 89004-300 89004-302 89004-324 89004-312 89004-304 89004-316
VWR® Microcentrifuge Tubes with Socket Screw-Cap
Tubes Microcentrifuge Tubes
These clear high grade gamma resistant PP tubes are ideal for sample processing applications.
  • Autoclavable at 121 ℃ and freezable to –80 ℃, and excellent for frozen storage
  • Caps accept color coded inserts, and easily write on the frosted side of insert
  • O-rings are made from silicone to insure a leakproof seal
  • RNase-, DNase-, and endotoxin-free
  • Sterile tubes are sterilized by irradiation
Excellent for freezer storage or secure sample handling. Screw caps incorporate silicone O-rings for a leakproof seal and accept color-coded inserts. Inserts snap into the top of the cap and create a flush surface. Tubes are available with standard caps or loop caps, which tether the cap to the tube.
Certifications: Manufactured in a Class 100,000 cleanroom environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management system. Meet the requirements of RoHS.
Ordering information: Additional screw caps and inserts are available in a variety of colors. Assorted pack 89004-362 and assorted case 89004-344 contain 100 of each: yellow, orange, red, blue, and green.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty