Would you like to log in again?
Monoject™ Sharps Containers, Covidien
Supplier: Covidien
Monoject™
8881676434 8881676285 8881676236
MJ8881-676285CS 403.02 USD InStock
MJ8881-676285 MJ8881-676434 89004-416
Monoject™ Sharps Containers, Covidien
Bins Sharps Containers
Containers feature polished viewing windows on all four sides, a clear lid with extra-wide butterfly closure, and needle removal notches
Containers can be autoclaved or incinerated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty