Would you like to log in again?
VWR® Good Laboratory Practice Notebooks
Supplier: VWR
VWR®
818-0117 VWR120LGO 818-0116 VWR120LLO VWR072LLO VWR072LGO VWR216LGO VWR216LLO
89005-128EA 54.39 USD InStock
89005-128 89005-138 89005-136 89026-648 89005-134 89026-646 89005-132 89005-130
VWR® Good Laboratory Practice Notebooks
Notebooks
VWR® Good Laboratory Practice Notebooks feature user data, guidelines, table of contents, and documentation pages with researcher and witness signature blocks.
  • Tamper-evident section-sewn binding
  • Acid-free archival safe paper
  • Title, project, and book number fields
  • Choose either 5mm grid or 5mm ruled page formats
  • Case bound in imitation leather cover specifically coated for durability in lab environments
Notebooks 89026-646 and -648 are oversized to allow attachment of full-size pages with room for signatures and dates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty