Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Reclosable UV-Protectant Amber Bags
Supplier: VWR
VWR®
MGUV3P0203 MGUV3P0305 MGUV3P0808 MGUV3P0608 MGUV3P0814 MGUV3P02509 MGUV3P0508 MGUV3P0406 MGUV3P1212
89005-326CS 275.87 USD InStock
89005-326 89005-324 89005-334 89005-322 89005-332 89005-320 89005-330 89005-318 89005-328
VWR® Reclosable UV-Protectant Amber Bags
Bags Storage Bags
Ideal for storage and transport of light-sensitive products or medications.
  • Re-closable design
  • Amber tint protects against UV radiation
  • Easy to identify with amber tint
Re-closable 3 mil bags feature a wide, white zipper to allow for repeated use and provides protection against dust and moisture. Amber tint makes bags easily identifiable.
Certifications: Bags meet light transmission rates between the ranges of full shade at DIN 53775 and DIN 53387, as well as reduced shade at DIN 53375.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty