Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trespa® Athlon® Shelves, AGR Fabricators

Supplier: AGR Fabricators

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Trespa® Trespa® Athlon®
AGS-4812-TR AGS-4818-TR AGS-4814-TR AGS-7214-TR AGS-4418-TR AGS-3614-TR AGS-4410-TR
89005-500EA 349.9 USD InStock
89005-500 89005-510 89005-498 89005-496 89005-494 89005-492 89005-506
Trespa® Athlon® Shelves, AGR Fabricators
Furniture Shelves
These phenolic resin shelves feature excellent chemical resistance.

  • Highly resistant to abrasives and scratches

Trespa® Athlon® material is an ideal choice for durability and attractiveness. Edges require no special treatment and can be easily profiled, milled, and polished. Color: black.

Solid, require no laminating or edge banding, and have excellent screw-holding ability for mechanical attachment to bench frames or partition hardware. Shelves can be easily cleaned with a mild detergent solution.

Min. compressive strength: 24,000 psi
Min. modulus of elasticity: 1.5 million psi
Min. shear strength: 2000 psi

Certifications: ASTM E84 class/A (25)

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty