Would you like to log in again?
Trespa® Toplab® Countertops, AGR Fabricators
Supplier: AGR Fabricators
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Trespa® Trespa® Toplab®
AGT-6130-TR AGT-3724-TR AGT-4924-TR AGT-7330-TR AGT-3730-TR AGT-6124-TR AGT-7324-TR AGT-4930-TR
89005-514EA 974.18 USD InStock
89005-514 89005-502 89005-512 89005-488 89005-508 89005-490 89005-516 89005-504
Trespa® Toplab® Countertops, AGR Fabricators
Furniture Casework
These compression-molded countertops are composed of thermoset resins with cellulose fiber reinforcement for outstanding resistance to chemicals, heat, and stains.
  • The nonporous tops will not rot or rust and are resistant to fungus, bacteria, and UV rays
  • Durable tops can be routinely disinfected and do not require surface dressing
  • Ease of machinability permits subsequent field modifications for equipment, piping, and wiring
Curved and other non-standard shaped tops are also possible. Countertops have a black core and black surface. Thickness: 2.5 cm (1").
Ordering information: Surface colors including light gray, light beige, and white are also available; contact your VWR sales representative for more information.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty