Would you like to log in again?
Axygen® Conical-Bottom Centrifuge Tubes, Polypropylene, Corning
Supplier: Corning
Axygen®
SCT-50ML-500 SCT-15ML-25-S SCT-15ML-500 SCT-50ML-R-S SCT-50ML-25-S
89005-624CS 220.73 USD InStock
89005-624 89005-622 89005-588 89005-586 89005-592
Axygen® Conical-Bottom Centrifuge Tubes, Polypropylene, Corning
Tubes Centrifuge Tubes
Polypropylene tubes feature printed graduations and a writing area for sample identification.
  • Caps may be closed with one hand
  • Tubes are autoclavable
The 15 ml tubes can be centrifuged to 7500 g; the 50 ml tubes can be centrifuged to 9400 g.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty