Would you like to log in again?
BX™ Series Protective Eyewear, 3M™
Supplier: 3M
BX™
7000052795 7000052795 7000052796 7000052796
89006-545CS 106.56 USD InStock
CA89006-497 89006-545 89006-497 CA89006-545
BX™ Series Protective Eyewear, 3M™
Safety Glasses
BX™ series protective eyewear features a soft fully adjustable nose bridge, lens angle adjustment, and adjustable length temples for extended wear.
  • Nose bridge is cushioned for softness and comfort
  • Sleek, contoured design offers an attractive appearance along with functional eye protection
  • Impact-resistant polycarbonate lenses absorb 99.9% UV rays
The ability to customize the safety glasses to the wearer makes them more pleasant to use over extended time periods.
Eye protection is important to safeguard the user's eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. All 3M™ protective eyewear combines safety features with comfort enhancements.
These safety glasses are recommended for use in assembly and mechanical, chipping, chiseling, cleaning, drilling, grinding, machining, masonry, metal repair, painting, pouring/casting, power fastening, riveting, sanding, and sawing applications. Industries in which these safety glasses are commonly used include aviation, construction, food and beverage, general manufacturing, mining, oil and gas, primary metals, and transportation.
Certifications: ANSI Z87.1-2003. CSA Z94.3-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty