Would you like to log in again?
Lexa™ Dust GoggleGear™ Series Safety Goggles, 3M™ Company
Supplier: 3M
GoggleGear™
7100075727 7010045732
89006-630BX 250.04 USD InStock
89006-630 89006-628
Lexa™ Dust GoggleGear™ Series Safety Goggles, 3M™ Company
Goggles
Lexa™ Dust GoggleGear™ Series Safety Goggles help protect eyes from flying particles with a soft foam flange that seals the entire lens perimeter, reducing the chances for contaminants to reach the eyes.
  • Fully assembled eyewear supplied ready to use
  • Soft foam flange provides comfort and helps keep out particulates
  • Top and bottom vent channels allow airflow and help minimize fogging
  • High wrap design allows for a wide field of view
  • Polycarbonate lens absorbs 99.9% UV
These safety goggles are recommended for use in assembly and mechanical, chipping, chiseling, cleaning, drilling, grinding, machining, masonry, metal repair, painting, pouring/casting, power fastening, riveting, sanding, and sawing applications. Industries in which these goggles are commonly used include construction, general manufacturing, military tactical, mining, oil and gas, and transportation.
Eye protection is important to safeguard user's eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. Comfortable and attractive eyewear increases the usage of safety goggles on the job. All 3M™ protective eyewear combines safety features with comfort enhancements.
These goggles may be used with existing Lexa™ Protective Eyewear (sold separately). To ensure clearer vision, goggles include top and bottom vent channels to increase airflow and help minimize fogging. Wrap around lens provides a wide field of view to enhance sightlines. The soft flange, lightweight frame, and low profile design all combine to enhance comfort. An adjustable headband keeps them securely in place.
Certifications: Meets ANSI Z87.1-2010, CSA Z94.3-2007.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty