Would you like to log in again?
PolarSafe™ 100-Place Freezer Storage Boxes, Argos Technologies
Supplier: Argos Technologies
PolarSafe™
24452-89 24452-91 24452-86 24452-90 24452-87 24452-88
89006-920CS 1271.12 USD InStock
89006-920 89006-930 89006-926 89006-928 89006-922 89006-924
PolarSafe™ 100-Place Freezer Storage Boxes, Argos Technologies
Boxes Cryoboxes
Storage boxes are suitable for use with 0.5, 1.5, and 2.0mL microtubes, as well as most other brands of cryogenic vials
Polypropylene boxes are a few millimeters taller than other available types of storage boxes and can accommodate several sizes of cryogenic vials or tubes of similar diameters up to 53.5mm (23/32") high. Boxes can withstand temperatures from –80° to 121°C (–112 to 250°F) and, unlike cardboard boxes, will not degrade after repeated thaws. The lid has an alphanumeric grid reference to aid with sample identification. Matte surfaces on box sides are suitable for marking or labeling. The box base and lid have impressed rims for stacking. All box bases are natural color to allow visual inspection.
Ordering information: Assorted color pack includes one box of each lid color.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty