Would you like to log in again?
Prinzing® Bilingual Safety Lockout Center, Brady Worldwide®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3002 3013 3002 3013
89007-132EA 340.63 USD InStock
CA89007-132 89007-132 CA89007-128 89007-128
Prinzing® Bilingual Safety Lockout Center, Brady Worldwide®
Security Locks
This English/Spanish safety lockout center is made of tough acrylic plastic and provides ready access to locks, tags, lockout devices, and information
Kit components include six multi-color padlocks, two packages of 10 bilingual lockout tags with ties, five 1.5" multiple lockout devices, fifteen English Lockout Safety booklets, and ten Spanish Lockout Safety booklets.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty