Would you like to log in again?
Large Lockout Station (Station Only), Brady Worldwide®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
50994 50994
89007-238EA 675.78 USD InStock
CA89007-238 89007-238
Large Lockout Station (Station Only), Brady Worldwide®
Security Locks
The large lockout station with components provides everything needed for locking out all types of valves and plugs
Constructed of sturdy white polystyrene with predrilled holes for easy wall mounting. Pegboard-style holes for hooks are drilled into the back (6 hooks supplied standard). Built-in lock rail holds 15 to 20 locks depending on lock size. Station is part of Large Lockout Station Kits 99698, 99699, and 99697.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty