Would you like to log in again?
Safety Padlock, Brady®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
51344 51353 51353 51339 51345 51347 51344 51347 51346 51339 51345 51346
89007-298PK 210.95 USD InStock
CA89007-296 89007-298 89007-296 CA89007-294 89007-302 CA89007-300 89007-300 CA89007-298 CA89007-302 89007-294 CA89007-292 89007-292
Safety Padlock, Brady®
Security Locks
Brady® Safety Padlocks are the ultimate locks for a wide variety of safety and security needs.
  • Unique cylinder is insulated to protect workers from shocks
  • Tamper resistant
  • Ribbed design for easier gripping and handling
Unique cylinder is insulated to protect workers from shocks when key is inserted. They cylinder is 6-pin precision-machined for more unique key cuts and better tamper resistance. Reserved, paracentric keyway provides optimal security and the special design prohibits the key from being released until the padlock shackle is closed. Locks are compact and lightweight, and they resist impact, temperature, chemicals, and corrosion. Their nonconductive, nonsparking body features a ribbed design for easier gripping and handling.
Shackle clearance is 38.1mm (1½"), shackle diameter is 6.35mm (¼"). Lock body measures 44.45H x 38.1W x 20.32D mm (1¾ x 1½ x ⅘").
Ordering information: Locks come standard with one key and English, French, and Spanish language labels. Padlocks are available keyed differently. Custom keying and charting is available.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty