Would you like to log in again?
Large Universal Valve Lockouts, Brady Worldwide®
Supplier: Brady
51388 51392 51388 51392
89007-342EA 226.29 USD InStock
89007-342 89007-346 CA89007-342 CA89007-346
Large Universal Valve Lockouts, Brady Worldwide®
Security Locks
Versatile, rugged, easy-to-use, and lightweight, the universal valve lockouts are effective for a variety of standard valves
They can also be used to lock out large levers, T-handles, and hard to secure mechanical devices. New open-ended clamp fits over closed rings and wide handles. Constructed of industrial-grade steel and nylon for extra impact and chemical resistance. Cable attachments are available in two formats. Choose from 1/8" nonconductive nylon cable or 1/8" rustproof sheathed metal cable. Additional base clamping units, blocking arms, and cable attachments sold separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty