Would you like to log in again?
GlobalMark® Tape, B-581, Brady Worldwide®
Supplier: Brady
76702 76703 76698 76697 76699 76701 76697 76701 76698 76703 76704 76699 76700 76704 76700 76702
89007-702EA 691.24 USD InStock
CA89007-706 CA89007-700 89007-702 89007-700 CA89007-698 CA89007-704 89007-706 CA89007-702 89007-704 89007-694 CA89007-692 89007-692 CA89007-696 89007-698 CA89007-694 89007-696
GlobalMark® Tape, B-581, Brady Worldwide®
Tapes
GlobalMark tapes are easy to install and very durable
Brady B-581 indoor/outdoor marking film is a high gloss, durable, flexible, pressure sensitive adhesive product, designed for use in cartridge form with the Labelizer Plus printer, VersaPrinter Label and Sign Maker System, and the HandiMark Portable Label Maker, and the GlobalMark Industrial Label Maker. Minimum Application Temperature: 50°F (10°C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty