Would you like to log in again?
U-Channel Sign Posts, Brady®
Supplier: Brady
97206 97205 97209 97209 97205 97207 97208 97210 97208 97207 97204 97204 95047 95047 97210
89007-918EA 143.59 USD InStock
CA89007-918 CA89007-938 89007-918 CA89007-948 89007-944 89007-942 CA89007-940 89007-938 89007-948 CA89007-946 CA89007-944 89007-946 CA89007-950 89007-940 89007-950
U-Channel Sign Posts, Brady®
Signs
Sign posts are ideal for mounting and installing traffic, utility, and industrial signs
Standard U-channel posts have a baked-green enamel finish on hot-rolled, high-tensile strength steel, allowing for years of maintenance-free performance. These sign posts weigh 0.9kg/linear meter (2lbs./linear foot) and have 9.5mm (3/8") holes, 2.5cm (1") on center. To fasten signs onto standard U-channel posts, use the vandal-proof Tufnut fastener. The Tufnut fastener is an 8mm (5/16") plated steel nut that secures tightly to prevent sign theft. Complete Tufnut set includes 2 bolts, 2 washers, and 3 Tufnut fasteners.
Tuff-N-Light™ posts are made of a lightweight, composite material that is maintenance-free. They are available in closed or open profile styles. Closed profile posts are six times torsionally stronger than steel posts and are pointed for easy installation. Open profile posts are for signs up to 30.5x45.7cm (12x18") in size. To fasten signs onto Tuff-N-Light™ U-channel posts, use Nuloc I™ or Nuloc II™ fasteners. The Nuloc™ I nylon fastener is maintenance-free and can be applied and taken off easily without the use of tools. The Nuloc II™ nylon fastener features adjustable tightening and may be reused, or its slip wings can be cut off for permanent applications. Both Nuloc™ sets includes two stems and two clips.
Two types of sign posts are available: standard U-channel posts and Tuff-N-Light™ U-channel posts.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty