Would you like to log in again?
polyproLABS® Fume Hoods, Air Control
Supplier: Air Control
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
polyproLABS™
FH-61-6-PP 4LP-VLF-10PP AC-110-4230 2C-HLF-4-PP 2C-HLF-5-PP 2C-HLF-8-PP FH-61-4-PP 5200 FH-61-10-PP AC-110-4836 4F-55-6-PP 4F-55-8-PP 2C-HLF-6-PP 5300 4LP-VLF-8-PP 4LP-VLF-5-PP 4LP-VLF-6-PP 4F-55-5-PP AC-110-4830 4F-55-10-PP AC-110-3630 5305 AC-110-3030 FH-61-8-PP 2C-HLF-10-PP FH-61-5-PP 4F-55-4-PP 4LP-VLF-4-PP
89008-252EA 5581.52 USD InStock
89008-252 89008-230 89008-196 89008-232 89008-210 89008-212 89008-234 89008-674 89008-214 89008-236 89008-676 89008-250 89008-248 89008-226 89008-228 89008-240 89008-680 89008-242 89008-682 89008-220 89008-222 89008-244 89008-246 89008-224 89008-238 89008-678 89008-216 89008-218
polyproLABS® Fume Hoods, Air Control
Fume Hoods and Enclosures Laminar Flow Cabinets
Polypropylene fume hoods are designed for full operator protection and continuous, easy access to the hood interior
Hoods feature fluorescent lighting contained in fume-tight compartments with translucent polypropylene windows. Laminar-flow hoods use lighting designed not to disrupt the movement of clean air. Lighting provides greater than 100 candlepower and is factory prewired to NEC and NFRA code. A vertical safety sash provides a full view and is counterbalanced by two plastic-coated cables. Sash moves easily and closes to within 2.5cm (1") of work surface. Hoods feature an air bypass located at the upper front interior of the hood that opens as the sash is lowered, providing a near-constant exhaust volume. Pressure drop across the fume hood exhaust system is less than 0.5in.H2O at 100cfm.
All-polypropylene cabinetry has a flush surface design and features all-polypropylene hinges, screws, handles, and other hardware. Doors are solid polypropylene with smooth beveled edges. Cabinets have leakproof floors to contain spills of corrosive substances.
Non-rusting hoods are specifically suited for the most corrosive environments. Hoods are manufactured with no exposed metals and are constructed with 1cm (3/8") thick stress-relieved, acid-resistant white polypropylene. All joints are thermally welded to ensure strong, leakproof, flush unions. The work surface is constructed of 2.5cm (1") thick polypropylene and features a front lip to contain spills. Each hood contains an internal baffle system also constructed of polypropylene. Baffle system provides safe and efficient removal of fumes when the hood is properly connected to a preexisting exhaust system. A manual adjustment is provided on the baffle system to allow the operator to adjust the negative pressure in the hood environment. Hoods have exhausts located across and above the work surface, both with adjustable louvered shutters. Units have a top-mounted round exhaust duct collar.
Ordering information: Tabletop fume hoods do not require a base; a base must be provided for all other cabinets listed. Various accessories are available (see 89008-204 series). Other options including various thicknesses of polypropylene, rectangular exhaust duct connections, exhaust failure alarms, and custom fitting to some other polyproLABS base cabinets (see 89008-376 series) are available. For more information, contact your VWR sales representative.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty