Would you like to log in again?
Alere iCassette® Multi-Panel, CLIA-Waived Drug Test Kits, Alere™ Toxicology
Supplier: Alere Toxicology
iCassette®
I-DOA-1125 I-DOA-1155 I-DOA-1135 I-DOA-3155 I-DOA-1165 I-DOA-1145 I-DOA-2155
89423-382PK 154.97 USD InStock
89423-382 89423-384 89423-380 89171-326 89423-386 89423-378 89008-838
Alere iCassette® Multi-Panel, CLIA-Waived Drug Test Kits, Alere™ Toxicology
Drug Tests
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Pipette style drug screen uses three drops of urine per well to detect drug metabolites in urine, with results in 5 minutes.
Room temperature storage, long shelf-life. Pipette included. Collection cup not included.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI) and (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX), Ecstacy (MDMA), Tricyclic Antidepressents (TCA), and Marijuana (THC) in human urine at the following cutoff levels (ng/mL) unless otherwise noted in description: AMP (1000), BAR (300), BZO (300), BUP (10), COC (300), mAMP (1000), OPI (2000), MOP (300), MTD (300), OXY (100), PCP (25), PPX (300), MDMA (500), TCA (1000), and THC (50).
Ordering information: The iScreen® Control Kit (item #88004) can be used with the following iCassettes®. Kit includes two vials (5mL positive and 5mL negative external controls). Refrigerated, must be shipped next day (sold separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty