Would you like to log in again?
BOTTLE WIDE-MOUTH PP 8OZ PK12
Supplier: Bel-Art Products, a Part of SP
PRECISIONWARE* PP Wide Mouth Bottles^ 250ml boston rounds are ideal for storing and shipping liquids and dry products. Heavy knurled PP caps have a watertight valve-seal. Translucent walls. Chemical and corrosion resistant. Autoclavable to 121 C. Remove caps before autoclaving. Cap Size: 45mm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOTTLE WIDE-MOUTH PP 8OZ PK12 89008-906 F10632-0006 Case of 72 Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty