Would you like to log in again?
Digital Urine Specific Gravity Refractometer, Model UG-[alpha], ATAGO®
Supplier: Atago
3464
89009-084EA 2672.56 USD InStock
89009-084
Digital Urine Specific Gravity Refractometer, Model UG-[alpha], ATAGO®
Refractometers Handheld Refractometers
Model UG-[alpha] is a compact urine specific gravity refractometer with digital display
Simple operation involves placing sample onto prism and pressing start. Measurements will stream across the LCD display. The minimum scale indication displays up to the fourth decimal. The UG-[alpha] refractometer is equipped with Automatic Temperature Compensation to provide accurate measurements, regardless of environmental conditions.
Ordering information: Uses one 9V battery.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty