Would you like to log in again?
TS Meter-D Digital Handheld Refractometer, Reichert
Supplier: Reichert
13960000
89009-332EA 3031.51 USD InStock
89009-332
TS Meter-D Digital Handheld Refractometer, Reichert
Refractometers Handheld Refractometers
This easy-to-use handheld digital refractometer is designed to automatically read samples with precision comparable to high-end automatic laboratory instruments
The TS Meter-D refractometer is equipped with five built-in scales offering a wide reading range and the versatility to read any sample required, regardless of the application. The device offers direct reading of urine specific gravity, serum or plasma protein concentration, cupric sulfate specific gravity, refractive index (nD), and Brix, all of which are temperature corrected. Simple calibration is provided for use in any setting. User-friendly three-button fully automatic operation provides fast, accurate results. A stainless steel well and hardened glass sample measurement surface are designed to withstand years of use, and can be easily cleaned and readied for sample measurement. This battery operated unit is portable for use in any location.
The optional IR communication package includes a docking station that allows for reading log, calibration log, and custom scale transfer to any laboratory PC via RS-232-C serial port.
The fully automatic readings result in the elimination of shadowline interpretation and the need to monitor countdown timers.
Ordering information: Uses four AAA batteries.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty