Would you like to log in again?
GASKET 4 THERMO EPDM 3PORT
Supplier: GARLOCK HYGIENIC TECHNOLOGIES, LLC.
Gasket^ Tri-Clamp* Sanitary Smart Gasket*, Process Testing/Validation Thermocouple, 4inch, EPDM, 3 port, FDA, USP Class VI
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GASKET 4 THERMO EPDM 3PORT 89011-698 G-TH-400-E3 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty