Would you like to log in again?
Jackson Safety™ G60 PURPLE NITRILE® Cut Resistant Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
Jackson Safety™ PURPLE NITRILE®
97432 97431 97432 97431 97434 97430 97433 97434 97430 97433
89011-812CS 495.75 USD InStock
89011-812 CA89011-812 89011-810 89011-804 CA89011-810 CA89011-808 89011-808 89011-806 CA89011-804 CA89011-806
Jackson Safety™ G60 PURPLE NITRILE® Cut Resistant Gloves, Kimberly-Clark
Gloves
New and improved G60 PURPLE NITRILE® gloves provide two times the cut resistance while maintaining dexterity and comfort.
  • Ambidextrous
  • Ideal for metal fabrication, glass/sharp part handling, and automotive assembly
  • Flexible, nitrile-dotted palm for enhanced grip
  • Dyneema® fabric enhances breathability
More tapered coating on the fingers provide added dexterity.
CPPT: 1150 g.
Certifications: ANSI level 3.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty