Would you like to log in again?
Double-Blade Squeegees, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CLN255 CLN254
56620-768EA 59.15 USD InStock
56620-768 89012-112
Double-Blade Squeegees, New Pig
Mops
Chemical-resistant PVC blades leave floors free of liquid and streaks.
  • Solid, 60" handle keeps users a safe distance from spill
  • Non-sparking, easy-to-clean PVC won't rust or corrode
Two blades double the service life because the head flips, and the unused blade acts as a splashguard.
Ordering information: Chemical-resistant PVC blades and 60" handle are sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty