Would you like to log in again?
KIMBLE® KIMAX® GL45 Media/Storage Bottles, Graduated, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
14395-2000 14396-100 14395-5000 14396-500 14396-5000 14395-500 14395-100 14395-250 14396-1000 14395-10000 14396-2000 14395-01 14395-1000 14396-10000 14396-250
89012-138CS 241.54 USD InStock
89012-138 89012-128 89000-938 89012-130 89000-928 89012-140 89012-132 89012-134 89012-136 89000-930 89000-932 89000-934 10798-966 89000-936 89000-926
KIMBLE® KIMAX® GL45 Media/Storage Bottles, Graduated, DWK Life Sciences
Bottles Cell Culture Bottles
General-purpose bottles feature white enamel graduations and marking spots.
Wide, 30 mm Int.Ø openings are designed for easy filling. Replaceable, drip-free, clear polypropylene pour ring is incorporated into the bottle design. Bottles are available uncapped, or supplied with linerless GL45 polypropylene caps that are autoclavable to 140 °C (284 °F). Optional high-temperature caps and pour rings are autoclavable to 180 °C (356 °F).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty