Would you like to log in again?
VWR® Signature™ Hooded Coveralls made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR
VWR® Signature™
IC180SWHLG0025VD IC180SWHMD0025VD IC180SWHMD0025VD IC180SWH3X0025VD IC180SWHXL0025VD IC180SWH2X0025VD IC180SWH2X0025VD IC180SWHLG0025VD
89012-820CS 841.76 USD InStock
89012-820 CA89012-812 89012-812 89012-814 89012-816 CA89012-814 89012-818 CA89012-818
VWR® Signature™ Hooded Coveralls made with DuPont™ Tyvek® IsoClean® Material
Disposable Garments
Coveralls provide high levels of protection and durability while remaining comfortable. A unique flash spinning process creates an excellent barrier against dry particulates, nonhazardous liquids, and microorganisms.
  • Protective, durable, and comfortable
  • Excellent barrier against dry particulates, nonhazardous liquids, and microorganisms
  • Chemically and biologically inert, low linting, and static free until laundered
  • Serged seams, front zipper closure, elastic wrists, and attached hood
  • Available with elastic ankles, or with attached boots that feature polyethylene anti-skid soles
Antistatic coveralls are low-linting and chemically and biologically inert.
All coveralls feature serged seams, front zipper closure, elastic wrists, and attached hood. Available with elastic ankles, or with attached boots that feature polyethylene anti-skid soles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty