Would you like to log in again?
VWR® Signature™ Boot Covers made with DuPont™ Tyvek® Material
Supplier: VWR
VWR® Signature™
IC446SWHLGVD IC446SWHMDVD IC446SWHLGVD IC446SWHMDVD IC446SWHXLVD
89013-156CS 543.76 USD InStock
CA89013-156 89013-156 89013-154 CA89013-154 89013-158
VWR® Signature™ Boot Covers made with DuPont™ Tyvek® Material
Disposable Garments
Boot covers provide a high level of protection and comfort for all cleanroom applications
  • High level of protection and comfort for all cleanroom applications
  • Nonwoven material protects against dry particulates and nonhazardous liquids
  • Soles made from PVC, coarsely textured interior prevents slipping inside and outside the garment
  • 45.7cm (18") high upper with elastic at leg openings
  • Supplied with ankle ties
Boot covers feature a 45.7cm (18") high upper with elastic at the top of the leg openings. They are supplied with ankle ties.
The nonwoven material offers protection against dry particulates as well as nonhazardous liquids. All boot covers are equipped with Gripper™ soles for slip-resistant performance. These soles are made from PVC and are combined with a coarsely textured interior to prevent slipping inside the garment as well as outside.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty