Would you like to log in again?
Uvex Strategy® Goggles, Honeywell Safety
Supplier: Honeywell Safety
uvex strategy®
S3815 S3800 S3820 S3810 S3805
89013-232CS 1716.18 USD InStock
89013-232 89013-230 89013-228 89013-226 89013-224
Uvex Strategy® Goggles, Honeywell Safety
Goggles
Uvex StrategyUvextreme® goggles offer all-day comfort, exceptional fit, and reliable protection.
Goggles are available with four ventilation options to meet most work applications: direct vent, closed vent, indirect vent, and indirect vent with foam lining to protect against dust, sand, or small particle incursion.
The lightweight, flexible TPE body conforms to the face, ensuring a snug fit and maximum protection in the harshest working conditions. The optically perfected polycarbonate lens delivers excellent peripheral vision and fits comfortably over most prescription eyewear. Lenses offer 99.9% UV protection and feature UvextraUvextreme® AF coating to minimize fogging. Goggle headbands are available in standard fabric or neoprene for chemical applications.
Certifications: Goggles comply with ANSI Z87+ and CSA Z94.3 standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty