Would you like to log in again?
A700 Series Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety
A701 A705 A702 A700 A700 A705 A705 A703 A700 A704
89020-068EA 6.72 USD InStock
89020-068 89020-066 CA89020-064 CA89020-075 CA89020-065 89020-072 89020-070 89020-064 CA89020-074 89020-074
A700 Series Protective Eyewear, Honeywell Safety
Safety Glasses
These economic and sporty glasses provide a stylish fit for the everyday wearer. The lightweight design is equipped with sport temples and soft temple tip pads that offer a secure, wraparound fit.
  • Molded universal nose bridge provides added comfort throughout extended use
  • Nine-base polycarbonate hardcoated lenses provide a 180° field of distortion-free vision, and offer 99.9% UV protection
The A700 series is available in a variety of lens styles for both indoor and outdoor work applications. Model 89020-074 features Fog-Ban® treatment to reduce fogging in extreme conditions.
Certifications: All models comply with ANSI Z87+ and CSA Z94.3 standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty