These industrial strength, anti-fatigue diamond-plate mats are designed for comfort and multiple shift operation use
- Industrial strength
- Antifatigue
- Resists abrasion and most chemical spills
The SpongeCote™ mats feature a 1cm (3/8") thick Nitricell® sponge base and have an overall thickness of 1.4cm (9/16"). The UltraSoft mats feature a double-thick Nitricell® sponge base and have an overall thickness of 2.4cm (15/16").
The diamond-plate surface is designed for heavy-use production and lab areas, and resists abrasion and most chemical spills. The Nitricell® base is a nitrile composite that provides exceptional comfort, rebound, and long life.