Would you like to log in again?
Ultra-Tred ArmorCote Anti-Fatigue Mat, Wearwell®
Supplier: WEARWELL
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
440.38X3X5BYL 440.38X2X60BYL 440.38X4X60BYL 440.38X3X12BYL 440.38X3X60BYL 440.38X2X3BYL
89020-640RL 1923.37 USD InStock
89020-640 89020-634 89020-632 89020-638 89020-756 89020-636
Ultra-Tred ArmorCote Anti-Fatigue Mat, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Ultra-Tred ArmorCote mats feature a more durable urethane outer skin than standard blown PVC sponge products.
  • Economic comfort for medium traffic areas
  • Urethane coating for greater abrasion resistance
  • Safety rounded edges
The tough outer skin increases service life, especially in applications where abrasion can cause the mat to fail. Ultra-Tred ArmorCote works longer where other entry-level sponge mats fail. Ideally suited for use in dry environments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty