Would you like to log in again?
ErgoDeck Heavy Duty Modular Anti-Fatigue Flooring, Wearwell®
Supplier: Wearwell
ErgoDeck®
560.78X6X18BK 560.78X6X15X15BK-CS4 560.78X18X18BK 560.78X6X18BK-CS10 71541180090 562.78X18X18BK-CS10 560.78X18X18BK-CS10 560.78X6X18YL 71541151243 562.78X18X18BK
89021-770EA 263.61 USD InStock
89021-770 89021-364 89021-772 89021-332 89021-322 89021-366 89021-324 89021-776 89021-326 89021-778
ErgoDeck Heavy Duty Modular Anti-Fatigue Flooring, Wearwell®
Floor Mats Permanent Floor Mats
Heavy duty compound allows carts to roll easily, while the box-spring design provides structure.
  • Standing relief in wet or dry applications
  • Customize in 3" increments to fit any area or configuration
  • Positive Interlocking System prevents separation
  • Easily replace single pieces or sections as needed to reduce costs
  • 100% silicone-free PVC tiles provide maximum durability
A comfortable, ergonomic floor that will withstand heavy use. The patented LockSafe® Positive Interlocking System delivers secure connections. Perfect for lean manufacturing, tiles can be unlocked and repositioned as work centers change to fit customer needs. The optional 6" side safety ramps and corners feature countersunk holes for semi-permanent installation. Available in a solid and drainage version, ErgoDeck Heavy-Duty withstands heavy use and cart traffic.
Certifications: Meets ASTM F1677. Dry: COF = 0.65/Wet: COF = 0.42. Meets ASTM D575-91/4.84%.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty