Would you like to log in again?
Leakbuster™ Containers with Molded O-Ring Caps, Starplex Scientific
Supplier: Starplex Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B1202-1W-OR B1202-1W-OR WB902-SO-OR
89027-046CS 137.17 USD InStock
CA89021-938 89027-046 89021-938
Leakbuster™ Containers with Molded O-Ring Caps, Starplex Scientific
Sample Containers General Purpose Containers
This premium quality line of containers consists of 60 mL, 90 mL, wide 90 mL and 120 mL sizes for leakproof transportation
Each polypropylene container is lot numbered and has clear graduations. The wide 90 mL and 120 mL containers feature a QuickTurn™ cap closure for easy application. Sterile containers have an orange cap and tamper tab patient label. The patient identification labels meet current JCAHO standards. Non-sterile containers have a yellow cap and a non-tabbed patient label. The standard 90 mL container is also available in an opaque polypropylene for discrete transport. A space saver dispenser box of 20, 90 mL containers is convenient for laboratories and Doctor's offices. Ideal for Operating Room use, both the 90 mL and 120 mL containers are packaged in a sterile, clear plastic peel pouch.
Ordering information: Leakbuster containers are packaged 100 containers per case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty