Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitri-Knit™ Supported Nitrile Gloves, Honeywell Safety

Supplier: Honeywell Safety

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nitri-Knit™
NK803ESIN/8 NK803ES/9 NK803/8-H5 NK803IN/10 NK803/11 NK803ESIN/11 NK803/7 NK803/8-H5 NK803/10 NK803/9 NK803IN/7 NK803ESIN/10 NK803ESIN/9 NK803IN/9 NK803ES/7 NK803ESIN/7 NK803IN/8 NK803IN/7 NK803ES/10 NK803IN/10 NK803ES/7 NK803ES/8 NK803ES/11 NK803/10 NK803IN/9 NK803ES/10 NK803/9 NK803IN/11 NK803IN/11 NK803/11 NK803ES/8 NK803ES/11 NK803ES/9 NK803IN/8
89022-162CS 2162.66 USD InStock
89022-162 89022-160 57100-106 57100-104 89092-990 CA89092-984 CA11001-469 CA89092-986 CA89022-158 CA89092-988 CA89022-156 89022-148 CA89022-154 89022-146 89022-144 89022-150 CA89022-160 CA57100-106 CA57100-104 CA57100-102 CA57100-100 89140-170 CA89092-990 CA11001-527 89092-984 89092-986 11001-526 11001-468 89092-988 89022-158 89022-156 89022-154 89022-152 CA89022-162
Nitri-Knit™ Supported Nitrile Gloves, Honeywell Safety
Gloves
For use in a wide range of applications, including: automotive assembly and painting; chemical handling and processing; degreasing; dip tank cleanup; fishing industry processing; food processing; printing cleanup; and general refinery, solvent, and maintenance work.

  • High nitrile content offers superior resistance to cuts, snags, and punctures
  • Interlock knit liner adds strength and comfort
  • Treated with permanent antibacterial and anti-mildew agent
  • Color-coded stitching makes size identification fast and easy
  • Compliant with USDA and FDA CFR 21 food processing regulations
  • Length: 30 cm (12")
  • Superior resistance to cuts, snags, and punctures
  • Interlock knit liner adds strength and comfort
  • Treated with permanent antibacterial and anti-mildew agent
  • Color-coded stitching makes size identification fast and easy
  • Compliant with USDA and FDA CFR 21 food-processing regulations

High nitrile content offers superior resistance to cuts, snags, abrasions, and punctures. Gloves provide excellent performance when exposed to solvents, caustics, and animal fats. Gloves feature unique rough texture finish for excellent grip on oily, wet, or dry objects. The interlock knit liner provides added strength and comfort. The nitrile exterior is permanently bonded to the inner liner for added durability. The hand portion of the glove is treated with a permanent antibacterial and anti-mildew agent. The 61 cm (24") extended sleeve gloves provide arm-to-shoulder protection. Gloves contain color-coded stitching for easy identification. Latex-free.

Certifications: Gloves comply with USDA and FDA regulations CFR 21 for use in food processing.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty