Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Fiber Glass Reinforced Containers, Molded Fiber Glass Tray Company
Supplier: MFG Tray
5090085269 5040085269 5081085269 5060085269 5070085269
89022-816EA 67.69 USD InStock
89022-816 89022-822 89022-812 89022-820 89022-818
Fiber Glass Reinforced Containers, Molded Fiber Glass Tray Company
Trays/Dishes/Pots Laboratory Trays/Dishes
These containers possess superior physical properties (temperature rating, weight capacity, chemical resistance) over competing thermoplastic products.
  • Design saves space - nests when empty
  • Can be used as a drawer
  • Freezer safe
They are ideal for storage of small parts.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty