Would you like to log in again?
Stacking Container Dolly, MFG
Supplier: MFG Tray
8701485136
89022-912EA 335.27 USD InStock
89022-912
Stacking Container Dolly, MFG
Carts
This heavy-duty fiberglass dolly will easily transport the content of MFG Tray's durable Stacking containers
These dollies and containers are resistant to cutting oils, grease, and solvents. Easy to clean with hot water or steam using standard detergents. Ideal for forgings, machined parts, stampings, ceramics, rubbers, and other heavy or hot parts emerging from molds or processing machines. Perfect for conveying heavy stacks of work-in-process parts on the standard 3” casters. MFG Tray is a leading manufacturer of high-strength, glass-reinforced composite containers, trays, boxes, flats, and totes for material handling in the metalworking, plastics, and electronics assembly industries, as well as in food service, confectionery, and pharmaceutical processing. MFG Tray products combine outstanding performance features with long service life to ensure exceptional value. The material matters in material handling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty