Would you like to log in again?
Econo™ Stevens Scissors, Floor Grade, Sklar
Supplier: Sklar
21-160 21-161
89023-056PK 127.97 USD InStock
89023-056 89023-058
Econo™ Stevens Scissors, Floor Grade, Sklar
Scissors
Econo™ Stevens Tenotomy scissors are used for delicate dissection and cutting, in ophthalmologic, neurological, and plastic surgery procedures.
Tenotomy scissors have long handles and small, sharp blades with sharp or blunt wedge-shaped tips. The long handles allow the surgeon to have an adequate grip and control while dissecting or cutting in small or narrow areas. Stevens tenotomy scissors are available straight or curved and with sharp or blunt tips, in order to accommodate different procedures. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used more often for dissecting and cutting mucus membranes. Econo™ instruments are Sklar's floor-grade, disposable instruments.
Made in Pakistan, they offer the greatest utility at the lowest cost. Econo™ instruments eliminate the need for cleaning and reprocessing. Because they are disposable, Econo™ instruments also reduce the likelihood of cross-contamination. Econo™ instruments are available in bulk quantities.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty