Would you like to log in again?
Econo™ Lucae Ear Forceps, Sklar
Supplier: Sklar
Econo™
21-180
89023-066PK 117.26 USD InStock
89023-066
Econo™ Lucae Ear Forceps, Sklar
Forceps
Sklar's Econo™ Lucae bayonet ear forceps are thumb forceps most commonly used for intra-aural procedures
  • Reduce cross contamination
  • Available in bulk quantities
Econo™ instruments eliminate the need for cleaning and reprocessing. The offset bayonet handle provides an unobstructed view of confined surgical sites.
They may also be used for procedures in the nasal cavity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty