Would you like to log in again?
Econo™ Lister Bandage Scissors, Floor Grade, Sklar®
Supplier: Sklar
Econo™
21-222 21-245 21-226 21-231 21-237 21-241 21-243
89023-092PK 167.63 USD InStock
89023-092 89023-082 76192-394 76192-392 89023-090 89023-078 89023-086
Econo™ Lister Bandage Scissors, Floor Grade, Sklar®
Scissors
Econo™ Lister Bandage scissors are a versatile instrument with multiple uses.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The jaws of the Lister Bandage scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
Econo™ instruments are floor-grade, disposable instruments. Made in Pakistan, they eliminate the need for cleaning and reprocessing. They also reduce the likelihood of cross-contamination. Econo™ instruments are available in bulk quantities.
They are used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty